
Methyl-3-bromo-5-(trifluoromethyl)benzoate 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:187331-46-0
:
Formula:C₉H₆BrF₃O₂
MDL Number:MFCD11847338
:283.0420 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together