1-(4-Fluoro-2-nitrophenyl)piperidine-4-carboxylic acid
Catalog # APOSPC5329-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:149…150 °C
Storage Temperature:Ambient
CAS Number:648917-79-7
Formula:C₁₂H₁₃FN₂O₄
MDL Number:MFCD05664786