Order Entry
Netherlands
ContactUsLinkComponent
1-(2-Fluoro-4-nitrophenyl)piperidine-2-carboxylic acid
1-(2-Fluoro-4-nitrophenyl)piperidine-2-carboxylic acid
Catalog # APOSPC5323-500MG
Supplier:  Apollo Scientific
CAS Number:  
1-(2-Fluoro-4-nitrophenyl)piperidine-2-carboxylic acid
Catalog # APOSPC5323-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
Melting Pt:153…154 °C
Storage Temperature:Ambient
CAS Number:1214189-74-8
Formula:C₁₂H₁₃FN₂O₄