1-[4-(Trifluoromethyl)phenyl]cyclopropanecarboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:886366-13-8
:
Formula:C₁₁H₉F₃O₂
MDL Number:MFCD07374440
:230.19 g/mol
Storage Temperature:Ambient
Frequently Bought Together