Order Entry
Netherlands
ContactUsLinkComponent
1-Bromo-5-(difluoromethyl)-2-fluorobenzene 97%
1-Bromo-5-(difluoromethyl)-2-fluorobenzene 97%
Catalog # APOSPC520902-25G
Supplier:  Apollo Scientific
CAS Number:  
1-Bromo-5-(difluoromethyl)-2-fluorobenzene 97%
Catalog # APOSPC520902-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :225.01 g/mol
Storage Temperature:Ambient
CAS Number:886509-99-5
Formula:C₇H₄BrF₃
MDL Number:MFCD08276255