Order Entry
Netherlands
ContactUsLinkComponent
1-Chloro-2-(difluoromethyl)-4-(trifluoromethyl)benzene 95%
1-Chloro-2-(difluoromethyl)-4-(trifluoromethyl)benzene 95%
Catalog # APOSPC520708-5G
Supplier:  Apollo Scientific
CAS Number:  
1-Chloro-2-(difluoromethyl)-4-(trifluoromethyl)benzene 95%
Catalog # APOSPC520708-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :230.57 g/mol
Storage Temperature:Ambient
CAS Number:1214383-34-2
Formula:C₈H₄ClF₅
MDL Number:MFCD14698551