1-Fluoro-2-(2,2,2-trifluoroethyl)-4-(trifluoromethyl)benzene 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1099597-68-8
:
Formula:C₉H₅F₇
MDL Number:MFCD11226656
:246.13 g/mol
Storage Temperature:Ambient
Frequently Bought Together