1-Chloro-4-(1,1-difluoroethyl)-2-(trifluoromethyl)benzene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1138444-94-6
:
Formula:C₉H₆ClF₅
MDL Number:MFCD11845801
:244.59 g/mol
Storage Temperature:Ambient
Frequently Bought Together