
Methyl-4-(trifluoromethyl)benzoate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2967-66-0
:
Formula:C₉H₇F₃O₂
MDL Number:MFCD00042324
:204.15 g/mol
Boiling Pt:86…87 °C/10mmHg
Melting Pt:13…14 °C
Density:1.268 g/cm³ (25 °C)
Flash Pt:82 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together