
2-(Methylamino)-5-(trifluoromethyl)-1,3,4-thiadiazole 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:25366-22-7
:
Formula:C₄H₄F₃N₃S
MDL Number:MFCD00042321
:183.16 g/mol
Melting Pt:115…117 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together