
2-Bromo-6-(trifluoromethyl)nicotinaldehyde 85+%
Catalog # APOSPC50083-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:254.01 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1192263-89-0
Formula:C₇H₃BrF₃NO
MDL Number:MFCD13193090