


p-Tolyl trifluoromethanesulphonate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:29540-83-8
:
Formula:C₈H₇F₃O₃S
MDL Number:MFCD00192507
:240.20 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
:GHS06
:GHS08
Frequently Bought Together