
4-(Trifluoromethyl)-1H-pyrazole 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:52222-73-8
:
Formula:C₄H₃F₃N₂
MDL Number:MFCD11226572
:136.077 g/mol
Melting Pt:73…78 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together