
3-Chloro-2-ethynyl-5-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1135283-18-9
:
Formula:C₈H₃ClF₃N
MDL Number:MFCD11840989
:205.57 g/mol
Boiling Pt:24 °C/0.1mmHg
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together