
2-Methoxy-5-(trifluoromethoxy)benzyl bromide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:478484-53-6
:
Formula:C₉H₈BrF₃O₂
MDL Number:MFCD04115961
:285.06 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together