Methyl-3-chloro-4-fluorobenzoate
Catalog # APOSPC49189-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:188.59 g/mol
Storage Temperature:Ambient
CAS Number:234082-35-0
Formula:C₈H₆ClFO₂
MDL Number:MFCD06203675