
2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}acetohydrazide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:446276-13-7
:
Formula:C₈H₈ClF₃N₄O
MDL Number:MFCD00141298
:268.63 g/mol
Melting Pt:161…163 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together