
2-{1-Amino-2-[3-(trifluoromethyl)phenoxy]ethylidene}malononitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:343375-42-8
:
Formula:C₁₂H₈F₃N₃O
MDL Number:MFCD00139203
:267.21 g/mol
Melting Pt:205…207 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together