

2-Fluoro-5-(trifluoromethyl)phenylacetonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:220227-59-8
:
Formula:C₉H₅F₄N
MDL Number:MFCD00061180
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
:GHS07
:GHS08
Frequently Bought Together