3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine
Catalog # APOSPC4375-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Boiling Pt:50…55 °C/11mmHg
Melting Pt:16…20 °C
Density:1.524 g/cm³ (25 °C)
Flash Pt:51 °C
:199.53 g/mol
Storage Temperature:Ambient
CAS Number:72537-17-8
Formula:C₆H₂ClF₄N
MDL Number:MFCD00191918