5-(4-Fluorophenyl)cyclohexane-1,3-dione 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:55579-72-1
:
Formula:C₁₂H₁₁FO₂
MDL Number:MFCD00174037
:206.22 g/mol
Melting Pt:173…179 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together