
Methyl (3-chloro-5-(trifluoromethyl)pyridin-2-ylsulphanyl)acetate
Catalog # APOSPC401003-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:285.67 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1024368-23-7
Formula:C₉H₇ClF₃NO₂S
MDL Number:MFCD00245199