Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(Trifluoromethyl)-1H-pyrazole
3-(Trifluoromethyl)-1H-pyrazole
3-(Trifluoromethyl)-1H-pyrazole
3-(Trifluoromethyl)-1H-pyrazole
APOSPC400547-1G
:
Apollo Scientific
:
3-(Trifluoromethyl)-1H-pyrazole
APOSPC400547-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
136.08 g/mol
:
Ambient
:
20154-03-4
:
C₄H₃F₃N₂
:
MFCD00115018