
2-Bromo-3-fluorobenzoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:132715-69-6
:
Formula:C₇H₄BrFO₂
MDL Number:MFCD01569398
:219.01 g/mol
Melting Pt:158…160 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together