
Octafluoroadipoyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:336-06-1
:
Formula:C₆Cl₂F₈O₂
MDL Number:MFCD00054726
:326.96 g/mol
Boiling Pt:42…45 °C/2mmHg
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
Frequently Bought Together