
5-Fluoro-o-tolylisothiocyanate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175205-39-7
:
Formula:C₈H₆FNS
MDL Number:MFCD00046800
:167.20 g/mol
Boiling Pt:101 °C/20mmHg
Storage Temperature:Ambient
EINECS:223-167-3
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together