Order Entry
Netherlands
ContactUsLinkComponent
8-Fluoro-4-hydroxy-2-(trifluoromethyl)quinoline 97%
Irritant
8-Fluoro-4-hydroxy-2-(trifluoromethyl)quinoline 97%
Catalog # APOSPC3756Y-10G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
8-Fluoro-4-hydroxy-2-(trifluoromethyl)quinoline 97%
Catalog # APOSPC3756Y-10G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :231.15 g/mol
Melting Pt:110…115 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:31009-31-1
Formula:C₁₀H₅F₄NO
MDL Number:MFCD00276580