2-Fluorobenzyl isothiocyanate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:64382-80-5
:
Formula:C₈H₆FNS
MDL Number:MFCD00046817
:167.21 g/mol
Boiling Pt:130 °C/10mmHg
Density:1.22 g/cm³ (25 °C)
Storage Temperature:Refrigerator
Frequently Bought Together