
4-Fluorobenzenesulphonyl chloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:349-88-2
:
Formula:C₆H₄ClFO₂S
MDL Number:MFCD00007438
:194.61 g/mol
Boiling Pt:95…96 °C/2mmHg
Melting Pt:29…31 °C
Flash Pt:113 °C
Storage Temperature:Ambient
EINECS:206-493-0
UN:3261
ADR:8,II
:GHS05
Frequently Bought Together