Order Entry
Netherlands
ContactUsLinkComponent
4-Bromo-2-fluorobenzenesulphonyl chloride 98%
Corrosive
4-Bromo-2-fluorobenzenesulphonyl chloride 98%
Catalog # APOSPC3459-100G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
4-Bromo-2-fluorobenzenesulphonyl chloride 98%
Catalog # APOSPC3459-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :273.51 g/mol
Boiling Pt:84 °C/0.5mmHg
Melting Pt:63…66 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
 :GHS05
CAS Number:216159-03-4
Formula:C₆H₃BrClFO₂S
MDL Number:MFCD01090993