Order Entry
Netherlands
ContactUsLinkComponent
Methyl-4-hydroxy-6-(trifluoromethyl)thieno-[3,4-b]-pyridine-1-carboxylate 97%
Irritant
Methyl-4-hydroxy-6-(trifluoromethyl)thieno-[3,4-b]-pyridine-1-carboxylate 97%
Catalog # APOSPC3409-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl-4-hydroxy-6-(trifluoromethyl)thieno-[3,4-b]-pyridine-1-carboxylate 97%
Catalog # APOSPC3409-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :277.22 g/mol
Melting Pt:144…145 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:175203-39-1
Formula:C₁₀H₆F₃NO₃S
MDL Number:MFCD00173841