
Tris(1,1,1,3,3,3-hexafluoro-2-propyl) Phosphite
Catalog # APOSPC3407-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:532.07 g/mol
Boiling Pt:130 °C
Density:1.69 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
CAS Number:66470-81-3
Formula:C₉H₃F₁₈O₃P
MDL Number:MFCD00192561