
5-Chloro-2-fluorobenzenesulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:351003-49-1
:
Formula:C₆H₃Cl₂FO₂S
MDL Number:MFCD03094221
:229.06 g/mol
Melting Pt:41…44 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
Frequently Bought Together