
3-Fluoro-2-iodobenzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:387-48-4
:
Formula:C₇H₄FIO₂
MDL Number:MFCD07774249
:266.0100 g/mol
Melting Pt:152…154 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together