
3,3'-Bis(trifluoromethyl)benzophenone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1868-00-4
:
Formula:C₁₅H₈F₆O
MDL Number:MFCD00000390
:318.22 g/mol
Melting Pt:98…102 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together