Order Entry
Netherlands
ContactUsLinkComponent
2-Methyl-5-(pentafluorothio)benzoic acid 98%
2-Methyl-5-(pentafluorothio)benzoic acid 98%
Catalog # APOSPC302817-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Methyl-5-(pentafluorothio)benzoic acid 98%
Catalog # APOSPC302817-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :262.19 g/mol
Storage Temperature:Ambient
CAS Number:623943-54-4
Formula:C₈H₇F₅O₂S
MDL Number:MFCD16652477