Order Entry
Netherlands
ContactUsLinkComponent
3-Chloro-4-ethoxy-5-fluorobenzoic acid 98%
3-Chloro-4-ethoxy-5-fluorobenzoic acid 98%
Catalog # APOSPC302527-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Chloro-4-ethoxy-5-fluorobenzoic acid 98%
Catalog # APOSPC302527-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :218.61 g/mol
Storage Temperature:Ambient
Formula:C₉H₈ClFO₃
MDL Number:MFCD09832336