3-(4-Methoxy-3-(trifluoromethyl)phenyl)propionic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1005205-20-8
:
Formula:C₁₁H₁₁F₃O₃
MDL Number:MFCD11519317
:248.20 g/mol
Melting Pt:94…97 °C
Storage Temperature:Ambient
Frequently Bought Together