Order Entry
Netherlands
ContactUsLinkComponent
3-[2-Fluoro-5-methyl-4-(trifluoromethyl)phenyl]propionic acid 98%
3-[2-Fluoro-5-methyl-4-(trifluoromethyl)phenyl]propionic acid 98%
Catalog # APOSPC302500-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[2-Fluoro-5-methyl-4-(trifluoromethyl)phenyl]propionic acid 98%
Catalog # APOSPC302500-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :250.19 g/mol
Storage Temperature:Ambient
CAS Number:1323966-25-1
Formula:C₁₁H₁₀F₄O₂
MDL Number:MFCD19687202