4-Methoxy-3-(trifluoromethyl)phenylacetonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1000555-32-7
:
Formula:C₁₀H₈F₃NO
MDL Number:MFCD09832301
:215.18 g/mol
Melting Pt:50…53 °C
Storage Temperature:Ambient
Frequently Bought Together