4-Fluoro-2-(trifluoromethoxy)benzylamine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1241828-99-8
:
Formula:C₈H₇F₄NO
MDL Number:MFCD19687190
:209.14 g/mol
Storage Temperature:Ambient
UN:3267
ADR:8,III
Frequently Bought Together