
3'-Methoxy-5'-(trifluoromethyl)acetophenone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:916421-06-2
:
Formula:C₁₀H₉F₃O₂
MDL Number:MFCD09025417
:218.17 g/mol
Melting Pt:54…58 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together