
3-[4-Chloro-2-(trifluoromethyl)phenyl]propionic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
:
Formula:C₁₀H₈ClF₃O₂
MDL Number:MFCD09025363
:252.62 g/mol
Melting Pt:87…90 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together