
2-Methoxy-3-(trifluoromethyl)phenylacetonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1017778-81-2
:
Formula:C₁₀H₈F₃NO
MDL Number:MFCD09832316
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
:GHS07
Frequently Bought Together