
2-Chloro-6-fluoro-3-methoxybenzaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:149949-29-1
:
Formula:C₈H₆ClFO₂
MDL Number:MFCD04115941
Melting Pt:105…106 °C
:188.59 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together