4-Fluoro-3-methyl-5-(trifluoromethyl)phenylacetonitrile 97%
Catalog # APOSPC302217-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:217.16 g/mol
Storage Temperature:Ambient
UN:3276
ADR:6.1,III
CAS Number:1373920-75-2
Formula:C₁₀H₇F₄N
MDL Number:MFCD22201073