3-[2-Methyl-4-(trifluoromethoxy)phenyl]propionic acid
Catalog # APOSPC302188-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:248.2 g/mol
Storage Temperature:Ambient
CAS Number:1373921-13-1
Formula:C₁₁H₁₁F₃O₃
MDL Number:MFCD22201060