3-(4-Chloro-3-fluorophenyl)propionic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:881189-65-7
:
Formula:C₉H₈ClFO₂
MDL Number:MFCD04116058
:202.61 g/mol
Melting Pt:80…84 °C
Storage Temperature:Ambient
Frequently Bought Together