N-(3-Fluoro-2-methyl-6-nitrophenyl)acetamide
Catalog # APOSPC300886-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:212.18 g/mol
Storage Temperature:Ambient
CAS Number:156243-41-3
Formula:C₉H₉FN₂O₃
MDL Number:MFCD03787363