2-(4-Bromophenoxy)-6-(trifluoromethyl)pyridine
Catalog # APOSPC300753-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:318.1 g/mol
Storage Temperature:Ambient
CAS Number:1086376-15-9
Formula:C₁₂H₇BrF₃NO
MDL Number:MFCD10703540